Learn More
1,3-Diiodopropane, 98+%, stab. with copper
Description
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Stable, but light sensitive - store in the dark. Incompatible with strong bases, strong oxidizing agents.

Specifications
Specifications
Melting Point | -20°C |
Density | 2.565 |
Boiling Point | 224°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.6415 |
Quantity | 250 g |
Beilstein | 1731117 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 295.89 |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 210-993-4 |
RTECSNumber | TY1080000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.