Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(1,3-Dimethyl-1H-pyrazol-5-yl)methylamine, 97%, Thermo Scientific™
Click to view available options
:
Amber Glass Bottle; 1g
Specifications
Specifications
CAS | 499770-63-7 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Molecular Formula | C6H11N3 |
MDL Number | MFCD02682038 |
Quantity | 1g |
Synonym | 1,3-dimethyl-1h-pyrazol-5-yl methanamine, 1,3-dimethyl-1h-pyrazol-5-yl methylamine, 2,5-dimethylpyrazol-3-yl methanamine, 1,3-dimethyl-1h-pyrazole-5-yl methanamine, 1-1,3-dimethyl-1h-pyrazol-5-yl methanamine, c-2,5-dimethyl-2h-pyrazol-3-yl-methylamine, 1h-pyrazole-5-methanamine, 1,3-dimethyl, 1,3-dimethylpyrazol-5-yl methylamine, 1h-pyrazole-5-methanamine,1,3-dimethyl, 5-aminomethyl-1,3-dimethyl-1h-pyrazole |
InChI Key | DAOWQCXPMWGSBW-UHFFFAOYSA-N |
SMILES | CN1N=C(C)C=C1CN |
IUPAC Name | 1-(1,3-dimethyl-1H-pyrazol-5-yl)methanamine |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction