Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-2-imidazolidinone, 98%

Catalog No. p-117959
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle

CAS: 80-73-9 | C5H10N2O | 114.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-73-9
Molecular Formula C5H10N2O
Molecular Weight (g/mol) 114.15
MDL Number MFCD00003188
InChI Key CYSGHNMQYZDMIA-UHFFFAOYSA-N
Synonym 1,3-dimethyl-2-imidazolidinone, n,n'-dimethylethyleneurea, dimethyl imidazolidinone, n,n'-dimethylimidazolidinone, 1,3-dimethylimidazolidone, rhonite 1, dmeu, karbomos tsem, 2-imidazolidinone, 1,3-dimethyl, 1,3-dimethylethyleneurea
PubChem CID 6661
IUPAC Name 1,3-dimethylimidazolidin-2-one
SMILES CN1CCN(C)C1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.2°C
Density 1.0500g/mL
Boiling Point 225.5°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4710 to 1.4730
Quantity 100 g
Fieser 11,202; 15,145
Specific Gravity 1.05
Merck Index 15, 3273
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol, ethanol, 2-propanol, ether,, methylene chloride, dmf, acetone, ketone and, chloroform
Viscosity 1.944 mPa.s (25°C)
Formula Weight 114.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
EINECSNumber 201-304-8
RTECSNumber NJ0660000
TSCA TSCA
Recommended Storage may discolor during storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.