Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-5-pyrazolone 98.0+%, TCI America™
SDP

Catalog No. D2031100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2031100G 100 g
D203125G 25 g
D2031500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2031100G Supplier TCI America Supplier No. D2031100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2749-59-9
Molecular Formula C5H8N2O
Molecular Weight (g/mol) 112.132
MDL Number MFCD00067008
InChI Key NDELSWXIAJLWOU-UHFFFAOYSA-N
Synonym 2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one
PubChem CID 17673
IUPAC Name 2,5-dimethyl-4H-pyrazol-3-one
SMILES CC1=NN(C(=O)C1)C

Specifications

Melting Point 117°C
Quantity 100 g
Formula Weight 112.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Dimethyl-5-pyrazolone
EINECSNumber (5)-5771
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.