Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Dimethylaminophenyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. D126925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D126925G 25 g
D12695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D126925G Supplier TCI America Supplier No. D126925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5339-01-5
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00059658
InChI Key XYGWUVVZDWJPBW-UHFFFAOYNA-N
Synonym 3-Dimethylamino-alpha-methylbenzyl Alcohol, 1-(3-Dimethylaminophenyl)ethyl Alcohol
PubChem CID 220552
IUPAC Name 1-[3-(dimethylamino)phenyl]ethanol
SMILES CC(C1=CC(=CC=C1)N(C)C)O

Specifications

Color Yellow
Boiling Point 102°C
Quantity 25 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(3-Dimethylaminophenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.