Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%

Catalog No. p-4258974
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438840100 10 g Glass bottle
AC438840500 50 g Glass bottle
AC438842500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC438840100 Supplier Thermo Scientific Chemicals Supplier No. 438840100
Only null left
Add to Cart

CAS: 1892-57-5 | C8H17N3 | 155.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1892-57-5
Molecular Formula C8H17N3
Molecular Weight (g/mol) 155.24
InChI Key LMDZBCPBFSXMTL-UHFFFAOYSA-N
Synonym 1-3-dimethylaminopropyl-3-ethylcarbodiimide, edac, 1-ethyl-3-3-dimethylaminopropyl carbodiimide, n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine, unii-rj5ozg6i4a, edci, 1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl, n-3-dimethylaminopropyl-n'-ethylcarbodiimide, rj5ozg6i4a, ethyldimethylaminopropyl carbodiimide
PubChem CID 15908
IUPAC Name 3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine
SMILES CCN=C=NCCCN(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8850g/mL
Boiling Point 66.0°C to 68.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
Quantity 10 g
Specific Gravity 0.885
Merck Index 15,3555
Formula Weight 155.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 217-579-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.