1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%

Manufacturer: Alfa Aesar A1080703

Quantity 1g

 View more versions of this product

Catalog No. AAA1080703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25952-53-8
    Molecular Formula C8H17N3·HCl
    Linear Formula CH3CH2N=C=N(CH2)3N(CH3)2·HCl
    Formula Weight 191.7
    MDL Number MFCD00012503
    Beilstein 5764110
    Synonym EDAC; EDCI
    Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride

    Specifications

    Formula Weight 191.7
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈113°C
    Health Hazard 1 Warning
    Molecular Formula C8H17N3·HCl
    Linear Formula CH3CH2N=C=N(CH2)3N(CH3)2·HCl
    CAS 25952-53-8
    Synonym EDAC; EDCI
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Recommended Storage 2°C to 8°C
    TSCA TSCA
    MDL Number MFCD00012503
    Beilstein 5764110
    Quantity 1g
    RTECS Number FF2200000
    Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride
    Assay ≥98%

    Description

    Applications
    1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride is used as a carboxyl activating agent. It is used in peptide synthesis, 3 ft.-amino-3 ft.-deoxyadenosine-5 ft.-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

    Solubility
    Soluble in acetonitrile dichloromethane, dimethylformamide, and tetrahydrofuran. Slightly soluble in water.

    Notes
    Sensitive to moisture. Incompatible with strong acids, strong oxidizing agents and moisture.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit