Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
22572-40-3
C9H20IN3
297.18
MFCD00011833
JHMDMCSGQQLHNA-UHFFFAOYSA-N
3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium,1-3-trimethylaminio propyl-3-ethylcarbodiimide,3-ethyliminomethylideneamino propyl-trimethylazanium,1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide,1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound,3-ethylimino methylidene amino propyl trimethylazanium,1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
122243
3-(ethyliminomethylideneamino)propyl-trimethylazanium
CCN=C=NCCC[N+](C)(C)C
22572-40-3
C9H20IN3
297.18
MFCD00011833
JHMDMCSGQQLHNA-UHFFFAOYSA-N
3-ethylimino methylidene amino-n,n,n-trimethylpropan-1-aminium,1-3-trimethylaminio propyl-3-ethylcarbodiimide,3-ethyliminomethylideneamino propyl-trimethylazanium,1-3-dimethylaminopropyl-3-ethylcarbodiimide methiodide,1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound,3-ethylimino methylidene amino propyl trimethylazanium,1-3-dimethylaminopropyl-3-ethylcarbodiimide, polymer-bound, 200-400 mesh, 2 % cross-linked with divinylbenzene
122243
3-(ethyliminomethylideneamino)propyl-trimethylazanium
CCN=C=NCCC[N+](C)(C)C
Coupling reagent
Melting Point 92.0°C to 97.0°C
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Merck Index 15, 3555
Solubility Information Solubility in water: 50g/L
Formula Weight 297.18
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respir

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to d

GHS Signal Word: Danger

EINECSNumber : 245-095-1

TSCA : TSCA