Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethylimidazolium Dimethyl Phosphate 97.0+%, TCI America™
SDP

Catalog No. D324025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D324025G 25 g
D32405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D324025G Supplier TCI America Supplier No. D324025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 654058-04-5
Molecular Formula C7H15N2O4P
Molecular Weight (g/mol) 222.181
InChI Key GSGLHYXFTXGIAQ-UHFFFAOYSA-M
Synonym 1,3-dimethylimidazolium dimethyl phosphate, acmc-1b6nn, 1,3-dimethylimidazoliumdimethylphosphate, dsstox_cid_27946, dsstox_rid_82698, dsstox_gsid_47970, 1-methyl-3-methylimidazolium dimethylphosphate, 1,3-dimethyl-imidazolium dimethylphosphate, 1,3-dimethylimidazolium dimethyl phosphate hplc
PubChem CID 16069365
IUPAC Name 1,3-dimethylimidazol-1-ium;dimethyl phosphate
SMILES CN1C=C[N+](=C1)C.COP(=O)([O-])OC

Specifications

Color Yellow
Quantity 25 g
Formula Weight 222.18
Percent Purity ≥97.0% (N)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethylimidazolium Dimethyl Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.