Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dioxolane, 99.5+%, pure, stabilized

Catalog No. p-3736365
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 646-06-0 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-06-0, 128-37-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -95°C
Color White
Density 1.0600g/mL
Boiling Point 74.0°C to 75.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.3990 to 1.4010
Quantity 1 L
Beilstein 19, 2
Fieser 14,281
Specific Gravity 1.06
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and acetone
Stabilizer 75ppm BHT max.
Viscosity 0.66 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 99.5+%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Dioxolane, Stabilized
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
missing translation for 'healthHazard3' GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear eye protection/face protection.
Wash f
EINECSNumber 211-463-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.