Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diphenoxybenzene 98.0+%, TCI America™
SDP

Catalog No. D266225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D266225G 25 g
D26625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D266225G Supplier TCI America Supplier No. D266225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3379-38-2
Molecular Formula C18H14O2
Molecular Weight (g/mol) 262.31
MDL Number MFCD00003036
InChI Key JTNRGGLCSLZOOQ-UHFFFAOYSA-N
Synonym m-diphenoxybenzene, benzene, 1,3-diphenoxy, m-diphenyloxybenzene, m-phenoxyphenoxybenzene, benzene, m-diphenoxy, resorcinol diphenyl ether, 1,1'-1,3-phenylenebis oxy dibenzene, m-diphenoxybenzol, acmc-1adxk
PubChem CID 76911
ChEBI CHEBI:39273
IUPAC Name 1,3-diphenoxybenzene
SMILES O(C1=CC=CC=C1)C1=CC(OC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 189°C
Quantity 25 g
Formula Weight 262.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.