Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diphenyl-1,3-propanedione, 98+%

Catalog No. aaa1345014
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 120-46-7 | C15H12O2 | 224.259 g/mol

1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diphenyl-1,3-propanedione, is a minor constituent of licorice found to inhibit mammary tumorigenesis, lymphomas, and leukemias in mice. It acts as a heat stabilizer& antitumour agent. It is used in perfumery and as well as in organic synthesis.

Solubility
It is Soluble in ether, chloroform, and aqueous sodium hydroxide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Acid anhydrides, Halogens, halogen acids. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00003085
InChI Key NZZIMKJIVMHWJC-UHFFFAOYSA-N
Synonym dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
PubChem CID 8433
ChEBI CHEBI:75417
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2

Specifications

Melting Point 76°C to 80°C
Boiling Point 219°C to 221°C (18 mmHg)
Quantity 25 g
Beilstein 514910
Merck Index 14,3009
Solubility Information It is Soluble in ether,chloroform,and aqueous sodium hydroxide. Insoluble in water.
Formula Weight 224.26
Percent Purity ≥98%
Chemical Name or Material 1,3-Diphenyl-1,3-propanedione
EINECSNumber 204-398-9
RTECSNumber TZ1930000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.