Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dithiane, 97%

Catalog No. p-3778466
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 505-23-7 | C4H8S2 | 120.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 505-23-7
Molecular Formula C4H8S2
Molecular Weight (g/mol) 120.23
MDL Number MFCD00006654
InChI Key WQADWIOXOXRPLN-UHFFFAOYSA-N
Synonym 1,3-dithiacyclohexane, m-dithiane, dithiane-1,3, 1,3-dithian, dithiane-1,3 french, 1,3 dithiane, ccris 6777, m-dithiane, 8ci, 1,3 dithiocyclohexane, acmc-209knk
PubChem CID 10451
ChEBI CHEBI:47845
IUPAC Name 1,3-dithiane
SMILES C1CSCSC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 49°C to 54°C
Color White to Beige
Density 90g/mL
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Fieser 02,182; 03,135; 04,216; 05,287; 06,248; 07,142; 14,157; 16,161; 01,208; 02,108; 12,158; 15,112
Formula Weight 120.23
Percent Purity 97%
Physical Form Crystals, Needles or Powder
Chemical Name or Material 1, 3-Dithiane
Show More Show Less
EINECSNumber 208-006-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.