Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dithiolane-2,4,5-trithione, oligomer, Thermo Scientific™

Catalog No. AC333120010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123399-69-9
Molecular Formula C3S5
Molecular Weight (g/mol) 196.333
MDL Number MFCD01632598
InChI Key JUQVDMLKRDJLKQ-UHFFFAOYSA-N
Synonym acmc-20kx1c, 1,3-dithiol-2,4,5-trithione, 1,3-dithiole-2,4,5-trithione
PubChem CID 1988934
IUPAC Name 1,3-dithiolane-2,4,5-trithione
SMILES C1(=S)C(=S)SC(=S)S1

Specifications

Color Yellow to Orange
Infrared Spectrum Authentic
Quantity 1 g
Formula Weight 196.35
Physical Form Powder
Chemical Name or Material 1,3-Dithiolane-2,4,5-trithione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.