Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Divinyl-1,1,3,3-tetramethyldisilazane 95.0+%, TCI America™
SDP

Catalog No. D176925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D176925ML 25 mL
D17695ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D176925ML Supplier TCI America Supplier No. D176925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7691-02-3
Molecular Formula C8H19NSi2
Molecular Weight (g/mol) 185.42
MDL Number MFCD00042846
InChI Key WYUIWUCVZCRTRH-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyl-1,3-divinyldisilazane, 1,3-divinyltetramethyldisilazane, 1,3-divinyl-1,1,3,3-tetramethyldisilazane, silanamine, 1-ethenyl-n-ethenyldimethylsilyl-1,1-dimethyl, bis dimethyl vinyl silyl amine, ccris 2450, bis ethenyldimethylsilyl amine, ethenyl-dimethylsilyl amino-dimethylsilyl ethene, n-dimethylvinylsilyl-1,1-dimethyl-1-vinylsilylamine, dvtmds
PubChem CID 82126
IUPAC Name [[[ethenyl(dimethyl)silyl]amino]-dimethylsilyl]ethene
SMILES C[Si](C)(C=C)N[Si](C)(C)C=C

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 mL
UN Number 2920
Formula Weight 185.42
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Divinyl-1,1,3,3-tetramethyldisilazane
RTECSNumber VV1302250
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.