Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Methoxyphenyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. M213525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M213525G 25 g
M21355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M213525G Supplier TCI America Supplier No. M213525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16015-71-7
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00040733
InChI Key PZIBVWUXWNYTNL-UHFFFAOYSA-N
Synonym 1-3-methoxyphenyl piperazine, n-3-methoxyphenyl piperazine, 1-3-methoxyphenyl-piperazine, unii-6lcp52962g, 1-3-methoxy-phenyl-piperazine, chembl59597, piperazine, 1-3-methoxyphenyl, 3-methoxy-1-piperazinylbenzene, pubchem8585, 3-meopp
PubChem CID 81430
IUPAC Name 1-(3-methoxyphenyl)piperazine
SMILES COC1=CC=CC(=C1)N2CCNCC2

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 g
UN Number 3267
Formula Weight 192.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-(3-Methoxyphenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.