Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Propanedithiol, 98%

Catalog No. p-3737258
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 109-80-8 | C3H8S2 | 108.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-80-8
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.22
MDL Number MFCD00004904
InChI Key ZJLMKPKYJBQJNH-UHFFFAOYSA-N
Synonym 1,3-propanedithiol, 1,3-dimercaptopropane, trimethylene dimercaptan, trimethylenedithiol, dithiotrimethyleneglycol, trimethylenedithioglycol, 1,3-propanedimercaptan, unii-r4luj82u52, fema no. 3588, 1,3-propane dithiol
PubChem CID 8013
ChEBI CHEBI:44864
IUPAC Name propane-1,3-dithiol
SMILES C(CS)CS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -79°C
Color Yellow
Density 1.0700g/mL
Boiling Point 169°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5380 to 1.542
Linear Formula HS(CH2)3SH
Quantity 50 mL
Beilstein 01, 476
Fieser 01,956; 04,413; 06,493; 09,394; 13,261
Specific Gravity 1.07
Merck Index 15, 7915
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol,ether,chloroform and,benzene
Formula Weight 108.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 3-Propanedithiol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
EINECSNumber 203-706-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.