Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Sulfopropyl)pyridinium Hydroxide Inner Salt 98.0+%, TCI America™
SDP

Catalog No. S081325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S081325G 25 g
S08135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S081325G Supplier TCI America Supplier No. S081325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15471-17-7
Molecular Formula C8H11NO3S
Molecular Weight (g/mol) 201.24
MDL Number MFCD00064468
InChI Key REEBJQTUIJTGAL-UHFFFAOYSA-N
Synonym 3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium
PubChem CID 84929
IUPAC Name 1-(3-sulfonatopropyl)pyridin-1-ium
SMILES [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1

Specifications

Melting Point 275°C
Color White
Quantity 25 g
Formula Weight 201.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Sulfopropyl)pyridinium Hydroxide Inner Salt [for Biochemical Research]
EINECSNumber (5)-3704&(5)-0758
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.