Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[3-(Trifluoromethyl)phenyl]ethanol, 98%, Thermo Scientific™

Catalog No. AAH3292203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3292203 1 g
AAH3292206 5 g
AAH3292214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3292203 Supplier Thermo Scientific Chemicals Supplier No. H3292203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 454-91-1
Molecular Formula C9H9F3O
Molecular Weight (g/mol) 190.17
MDL Number MFCD00021868
InChI Key YNVXCOKNHXMBQC-UHFFFAOYNA-N
Synonym 1-3-trifluoromethyl phenyl ethanol, alpha-methyl-3-trifluoromethyl benzyl alcohol, 1-3-trifluoromethyl phenyl ethan-1-ol, alpha-methyl-3-trifluoromethylbenzyl alcohol, pubchem7467, .alpha.-methyl-m-trifluoromethylbenzyl alcohol, 3-trifluoromethylphenylethanol, 3-trifluoromethylphenyl ethanol, 1-3-trifluoromethyl-phenyl ethanol
PubChem CID 136296
IUPAC Name 1-[3-(trifluoromethyl)phenyl]ethanol
SMILES CC(O)C1=CC=CC(=C1)C(F)(F)F

Specifications

Density 1.249
Boiling Point 100°C to 102°C (17 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.458
Quantity 1 g
Formula Weight 190.17
Percent Purity 98%
Chemical Name or Material 1-[3-(Trifluoromethyl)phenyl]ethanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.