Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,7,10-Tetraazacyclododecane Tetrahydrochloride 98.0+%, TCI America™
SDP

Catalog No. p-7136769 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 10045-25-7
Molecular Formula C8H24Cl4N4
Molecular Weight (g/mol) 318.108
MDL Number MFCD00068484
InChI Key SBWLCGZEBQGYRP-UHFFFAOYSA-N
Synonym 1,4,7,10-tetraazacyclododecane tetrahydrochloride, cyclen tetrahydrochloride, cyclen-4hcl, unii-51830y7tix, tetraaza-12-crown-4 tetrahydrochloride, 1,4,7,10-tetraazacyclododecane, tetrahydrochloride, 1,4,7,10-tetraazacyclododecane, hydrochloride 1:4, 1,4,7,10-tetraazacyclododecanetetrahydrochloride, cyclen tetrahydrochloride mi, 1,4,7,10-tetraazacyclododecane, chloride, chloride, chloride, chloride
PubChem CID 11652630
IUPAC Name 1,4,7,10-tetrazacyclododecane;tetrahydrochloride
SMILES C1CNCCNCCNCCN1.Cl.Cl.Cl.Cl

Specifications

Melting Point 280°C
Color White-Yellow
Quantity 1 g
Formula Weight 318.11
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4,7,10-Tetraazacyclododecane Tetrahydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.