Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,4,7,10-Tetraazacyclotridecane
Click to view available options
Quantity:
250 mg
1 g
Description
1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.
Solubility
Soluble in water.
Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1,4,7,10-Tetraazacyclotridecane is used to find the binding constants, and for building block elements.
Solubility
Soluble in water.
Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Quantity | 1 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 186.3 |
Chemical Name or Material | 1,4,7,10-Tetraazacyclotridecane |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction