Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,7-Triazacyclononane trihydrochloride, 97%

Catalog No. AC294310025
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle

CAS: 58966-93-1 | C6H18N3 | 132.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58966-93-1
Molecular Formula C6H18N3
Molecular Weight (g/mol) 132.23
MDL Number MFCD00074887
InChI Key ITWBWJFEJCHKSN-UHFFFAOYSA-Q
Synonym 1,4,7-triazonane trihydrochloride, 1,4,7-triazacyclononane trihydrochloride, 1,4,7-triazacyclononane tri hydrochloride, octahydro-1h-1,4,7-triazonine trihydrochloride, 1h-1,4,7-triazonine, octahydro-, trihydrochloride, pubchem21332, 1,4,7-triazacyclononanetrihydrochloride, acmc-1au73, c6h15n3.3hcl, ksc911s7j
PubChem CID 2724990
SMILES C1C[NH2+]CC[NH2+]CC[NH2+]1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 285.0°C to 290.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 2.5 g
IUPAC Name 1,4,7-triazonane;trihydrochloride
Formula Weight 238.59
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1, 4, 7-Triazacyclononane trihydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.