Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4,8,11-Tetraazacyclotetradecane, 98%

Catalog No. ac269330010
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 295-37-4 | C10H24N4 | 200.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 295-37-4
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
MDL Number MFCD00005105
InChI Key MDAXKAUIABOHTD-UHFFFAOYSA-N
Synonym 1,4,8,11-tetraazacyclotetradecane, cyclam, 14 anen4, 1,4,8,11-tetraazacyclo-tetradecane, 1,4,8,11-tetraazacyclotetradecane cyclam, ips2, acmc-209h9n, 5-26-11-00027 beilstein handbook reference, 1,8,11-tetraazacyclotetradecane, 1,4,8,11tetraaza-cyclotetradecane
PubChem CID 64964
ChEBI CHEBI:37401
IUPAC Name 1,4,8,11-tetrazacyclotetradecane
SMILES C1CNCCNCCCNCCNC1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 185.0°C to 188.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 26,1647
Fieser 17,89
Solubility Information Solubility in water: 5g/100mL (20°C).
Formula Weight 200.33
Percent Purity 98%
Physical Form Fluffy Needle-like Crystalline Powder
Chemical Name or Material 1,4,8,11-Tetraazacyclotetradecane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 206-039-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.