Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Acetylpiperidino)ethan-1-one, 97%, Thermo Scientific™

Catalog No. cd11879cb
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 162368-01-6
Molecular Formula C9H15NO2
Molecular Weight (g/mol) 169.22
MDL Number MFCD00106774
InChI Key VFZXZEJGLKOTBW-UHFFFAOYSA-N
Synonym 1-4-acetylpiperidino ethan-1-one, 1-1-acetylpiperidin-4-yl ethanone, 1,4-diacetylpiperidine, 1-4-acetylpiperidin-1-yl ethan-1-one, 1,1'-piperidine-1,4-diyl diethanone, 1-1-acetyl-piperidin-4-yl-ethanone, piperidine, 1,4-diacetyl-9ci, 1-4-acetylpiperidin-1-yl ethanone, ethanone,1-1-acetyl-4-piperidinyl, piperidine, 1,4-diacetyl
PubChem CID 2777250
IUPAC Name 1-(1-acetylpiperidin-4-yl)ethanone
SMILES CC(=O)C1CCN(CC1)C(C)=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 169.22
Percent Purity 97%
Chemical Name or Material 1-(4-Acetylpiperidino)ethan-1-one
Hazard Category Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.