Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine 96.0+%, TCI America™
SDP

Catalog No. M192025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M192025G 25 g
M19205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M192025G Supplier TCI America Supplier No. M192025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74852-62-3
Molecular Formula C17H21N3O
Molecular Weight (g/mol) 283.375
MDL Number MFCD02071496
InChI Key VXEGSRKPIUDPQT-UHFFFAOYSA-N
Synonym 4-[4-(4-Methoxyphenyl)-1-piperazinyl]aniline
PubChem CID 154406
IUPAC Name 4-[4-(4-methoxyphenyl)piperazin-1-yl]aniline
SMILES COC1=CC=C(C=C1)N2CCN(CC2)C3=CC=C(C=C3)N

Specifications

Melting Point 186°C
Quantity 25 g
Formula Weight 283.38
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.