Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Benzenediboronic acid, 96%

Catalog No. AAB2406403
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. AAB2406403

Supplier: Thermo Scientific Chemicals B2406403

Only null left
Add to Cart

CAS: 4612-26-4 | C6H8B2O4 | 165.746 g/mol

It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for palladium-catalyzed sequential alkenylation and conjugate addition reactions, scholl cyclizations, Suzuki-Miyaura cross-coupling reactions. It is also used in the preparation of indolizine derivatives as OLEDs, in organic thin-film transistors, Fluorescence and solution-processable coordination polymers.

Solubility
Soluble in water 2.5%.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4612-26-4
Molecular Formula C6H8B2O4
Molecular Weight (g/mol) 165.746
MDL Number MFCD00236018
InChI Key BODYVHJTUHHINQ-UHFFFAOYSA-N
Synonym 1,4-benzenediboronic acid, 1,4-phenylenediboronic acid, 1,4-phenylenebisboronic acid, benzene-1,4-diboronic acid, 4-boronophenyl boronic acid, p-benzenediboronic acid, p-phenylenediboronic acid, 1,4-phenylenebis boronic acid, 1,4 phenyl diboronic acid, boronic acid, 1,4-phenylenebis
PubChem CID 230478
IUPAC Name (4-boronophenyl)boronic acid
SMILES B(C1=CC=C(C=C1)B(O)O)(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 2836921
Solubility Information Soluble in water 2.5%.
Formula Weight 165.75
Percent Purity 96%
Chemical Name or Material 1,4-Benzenediboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.