Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Benzenedimethanol, 99%

Catalog No. p-7021884
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 589-29-7 | C8H10O2 | 138.166 g/mol

1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.

Solubility
Soluble in ether, toluene, methanol and hot water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-29-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004665
InChI Key BWVAOONFBYYRHY-UHFFFAOYSA-N
Synonym 1,4-benzenedimethanol, p-xylylene glycol, p-xylene glycol, terephthalyl alcohol, 4-hydroxymethyl phenyl methanol, 1,4-xylylene glycol, 1,4-phenylenedimethanol, p-benzenedimethanol, p-xylylenediol, 1,4-dimethylolbenzene
PubChem CID 11506
IUPAC Name [4-(hydroxymethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CO)CO

Specifications

Melting Point 117°C to 121°C
Boiling Point 138°C to 143°C (1 mmHg)
Quantity 5 g
Beilstein 2042077
Solubility Information Soluble in ether,toluene,methanol and hot water.
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 1,4-Benzenedimethanol
EINECSNumber 209-641-2
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.