Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4'-Bipiperidine, 99%

Click to view available options
Quantity:
1g
5g
25g
4897-50-1
C10H20N2
168.284
MFCD00006475
QDVBKXJMLILLLB-UHFFFAOYSA-N
4-piperidinopiperidine,1,4'-bipiperidine,4-piperidino-piperidine,1,4'-bipiperidyl,4-1-piperidinyl piperidine,1,4' bipiperidinyl,4-piperidin-1-yl piperidine,1,4-bipiperidine,1-piperidin-4-yl piperidine,4-piperidin-1-ylpiperidine
78607
CHEBI:40117
1-piperidin-4-ylpiperidine
C1CCN(CC1)C2CCNCC2
4897-50-1
C10H20N2
168.284
MFCD00006475
QDVBKXJMLILLLB-UHFFFAOYSA-N
4-piperidinopiperidine,1,4'-bipiperidine,4-piperidino-piperidine,1,4'-bipiperidyl,4-1-piperidinyl piperidine,1,4' bipiperidinyl,4-piperidin-1-yl piperidine,1,4-bipiperidine,1-piperidin-4-yl piperidine,4-piperidin-1-ylpiperidine
78607
CHEBI:40117
1-piperidin-4-ylpiperidine
C1CCN(CC1)C2CCNCC2
Melting Point 66°C to 70°C
Quantity 1g
UN Number UN3259
Formula Weight 168.28
Percent Purity 99%
Chemical Name or Material 1,4'-Bipiperidine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.