Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. B442825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B442825G 25 g
B44285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B442825G Supplier TCI America Supplier No. B442825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94525-05-0
Molecular Formula C20H14F6N2O2
Molecular Weight (g/mol) 428.33
MDL Number MFCD07368071
InChI Key LACZRKUWKHQVKS-UHFFFAOYSA-N
PubChem CID 19805071
IUPAC Name 4-{4-[4-amino-2-(trifluoromethyl)phenoxy]phenoxy}-3-(trifluoromethyl)aniline
SMILES NC1=CC(=C(OC2=CC=C(OC3=C(C=C(N)C=C3)C(F)(F)F)C=C2)C=C1)C(F)(F)F

Specifications

Melting Point 134°C
Color White
Quantity 25 g
Formula Weight 428.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(4-amino-2-trifluoromethylphenoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.