Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(dimethylsilyl)benzene 98.0+%, TCI America™
SDP

Catalog No. p-7123721
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 2488-01-9
Molecular Formula C10H16Si2
Molecular Weight (g/mol) 192.41
MDL Number MFCD00039790
InChI Key UHXCHUWSQRLZJS-UHFFFAOYSA-N
Synonym 1,4-bis dimethylsilyl benzene, benzene, 1,4-bis dimethylsilyl, p-phenylenebis dimethylsilane, 1,4-phenylenebis dimethylsilane, p-bis dimethylsilyl benzene, silane, p-phenylenebis dimethyl, silane, 1,4-phenylenebis dimethyl, 1,4 bis dimethylsilyl benzene
PubChem CID 6328729
IUPAC Name (4-dimethylsilylidenecyclohexa-2,5-dien-1-ylidene)-dimethylsilane
SMILES C[Si](=C1C=CC(=[Si](C)C)C=C1)C

Specifications

Color Yellow
Boiling Point 220°C
Quantity 5 mL
Formula Weight 194.42
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Bis(dimethylsilyl)benzene
RTECSNumber VV4830000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.