Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(methoxydimethylsilyl)benzene, 97%

Catalog No. p-7030362
Click to view available options
Quantity:
1 g
5 g

CAS: 131182-68-8 | C12H22O2Si2 | 254.476 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131182-68-8
Molecular Formula C12H22O2Si2
Molecular Weight (g/mol) 254.476
MDL Number MFCD09266162
InChI Key VQECZZRUEDMVCU-UHFFFAOYSA-N
Synonym 1,4-bis methoxydimethylsilyl benzene, methoxy 4-methoxydimethylsilyl phenyl dimethylsilane, amtsi067, 1,4-phenylenebis methoxydimethylsilane, p-bis dimethylmethoxysilyl benzene, 1,4-phenylene bis methoxy dimethyl silane, methoxy-4-methoxy dimethyl silyl phenyl-dimethylsilane
PubChem CID 21900341
IUPAC Name methoxy-[4-[methoxy(dimethyl)silyl]phenyl]-dimethylsilane
SMILES CO[Si](C)(C)C1=CC=C(C=C1)[Si](C)(C)OC

Specifications

Quantity 1 g
Formula Weight 254.47
Percent Purity 97%
Sensitivity Moisture sensitive
Chemical Name or Material 1,4-Bis(methoxydimethylsilyl)benzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.