Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromobutoxy)-4-fluorobenzene, Thermo Scientific™

Catalog No. AC383100050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2033-80-9
Molecular Formula C10H12BrFO
Molecular Weight (g/mol) 247.107
MDL Number MFCD02030562
InChI Key SESKDKZVNDEQBM-UHFFFAOYSA-N
Synonym 1-4-bromobutoxy-4-fluorobenzene, benzene, 1-4-bromobutoxy-4-fluoro, 1-bromo-4-p-fluorophenoxy butane, 4-4-fluorophenyl oxybutyl bromide, 1-bromo-4-4'-fluorophenoxy butane, 1-bromo-4-4-fluorophenoxy butane
PubChem CID 7172295
IUPAC Name 1-(4-bromobutoxy)-4-fluorobenzene
SMILES C1=CC(=CC=C1OCCCCBr)F

Specifications

Packaging Glass Bottle
Linear Formula Br(CH2)4OC6H4F
Quantity 5 g
Formula Weight 247.11
Chemical Name or Material 1-(4-Bromobutoxy)-4-fluorobenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.