Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromobutoxy)-4-methylbenzene, 95%, Thermo Scientific™

Catalog No. AC379290050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3257-49-6
Molecular Formula C11H15BrO
Molecular Weight (g/mol) 243.144
MDL Number MFCD02030566
InChI Key WKFSNKTYOQGTBN-UHFFFAOYSA-N
Synonym 1-4-bromobutoxy-4-methylbenzene, toluene, p-4-bromobutoxy, benzene, 1-4-bromobutoxy-4-methyl, 4-methylphenoxybutylbromide, toluene, p-4-bromobutoxy-,, 4-4-methylphenoxy butyl bromide, 1-bromo-4-4-methylphenoxy butane
PubChem CID 137870
IUPAC Name 1-(4-bromobutoxy)-4-methylbenzene
SMILES CC1=CC=C(C=C1)OCCCCBr

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Refractive Index 1.5360 to 1.5380 (20°C, 589nm)
Linear Formula CH3C6H4O(CH2)4Br
Quantity 5 g
Formula Weight 243.14
Percent Purity ≥94%
Physical Form Liquid After Melting
Chemical Name or Material 1-(4-Bromobutoxy)-4-methylbenzene
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.