Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)-2-pyrrolidinone, 99+%, Thermo Scientific™

Catalog No. AC271310010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7661-32-7
Molecular Formula C10H10BrNO
Molecular Weight (g/mol) 240.1
MDL Number MFCD00138502
InChI Key YINFEFUSAQRZGG-UHFFFAOYSA-N
Synonym 1-4-bromophenyl pyrrolidin-2-one, 1-4-bromophenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-4-bromophenyl, 1-4-bromophenyl-2-pyrrolidone, pubchem9356, p-bromo-phenylpyrrolidone, acmc-209p4u, buttpark 32\07-61, n-4-bromophenyl-2-pyrrolidinone, 1-4-bromophenyl pyrrolidine-2-one
PubChem CID 736110
IUPAC Name 1-(4-bromophenyl)pyrrolidin-2-one
SMILES C1CC(=O)N(C1)C2=CC=C(C=C2)Br

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 98.0°C to 101.0°C
Color Beige or Pink
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 240.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Bromophenyl)-2-pyrrolidinone, 99+%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.