Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromophenyl)hexane, 97%, Maybridge™

Click to view available options
Quantity:
1g
10g
25g
23703-22-2
C12H17Br
241.17
MFCD00061114
MXHOLIARBWJKCR-UHFFFAOYSA-N
1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
598186
1-bromo-4-hexylbenzene
CCCCCCC1=CC=C(Br)C=C1
23703-22-2
C12H17Br
241.17
MFCD00061114
MXHOLIARBWJKCR-UHFFFAOYSA-N
1-4-bromophenyl hexane, 4-hexylbromobenzene, 1-bromo-4-n-hexylbenzene, 4-bromo-1-hexylbenzene, benzene, 1-bromo-4-hexyl, 4-bromo-n-hexylbenzene, 4-hexylphenyl bromide, 1-bromo-4-hexyl-benzene, acmc-1cb1g, #
598186
1-bromo-4-hexylbenzene
CCCCCCC1=CC=C(Br)C=C1
Quantity 25g
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H4(CH2)5CH3
Formula Weight 241.17
Percent Purity 97%
Chemical Name or Material 1-(4-Bromophenyl)hexane