Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals

Catalog No. AC154310025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC154310025 2.5 kg
AC154315000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154310025 Supplier Thermo Scientific Chemicals Supplier No. 154310025
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2425-79-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00005146
InChI Key SHKUUQIDMUMQQK-UHFFFAOYNA-N
Synonym 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
PubChem CID 17046
IUPAC Name 2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
SMILES C(CCOCC1CO1)COCC1CO1

Specifications

CAS Min % 50.0
Color Yellow
Density 1.1000g/mL
Boiling Point 266.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 55% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4650
Quantity 2.5 kg
Specific Gravity 1.1
Solubility Information Solubility in water: 55,6g/L (20°C).
Viscosity <15 mPa.s (25°C)
Formula Weight 202.25
Percent Purity 60%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1, 4-Butanediol diglycidyl ether, 60%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful to aquatic life with long l
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 219-371-7
RTECSNumber EJ5100000
TSCA TSCA
Recommended Storage This product unavoidably and progressively degrades on storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.