Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
72934-37-3
C10H9ClO2
196.63
MFCD00001289
YAHLWSGIQJATGG-UHFFFAOYSA-N
1-4-chlorophenyl cyclopropanecarboxylic acid,1-4-chlorophenyl cyclopropane-1-carboxylic acid,1-4-chlorophenyl-1-cyclopropanecarboxylic acid,cyclopropanecarboxylic acid, 1-4-chlorophenyl,1-p-chlorophenyl cyclopropanecarboxylic acid,acmc-209oos,1-4-chlorophenyl cyclopropanecarboxylicacid,1-4chloro-phenyl-cyclopropylcarboxylic acid,1-4-chloro-phenyl-cyclopropylcarboxylic acid
98606
1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
C1CC1(C2=CC=C(C=C2)Cl)C(=O)O
72934-37-3
C10H9ClO2
196.63
MFCD00001289
YAHLWSGIQJATGG-UHFFFAOYSA-N
1-4-chlorophenyl cyclopropanecarboxylic acid,1-4-chlorophenyl cyclopropane-1-carboxylic acid,1-4-chlorophenyl-1-cyclopropanecarboxylic acid,cyclopropanecarboxylic acid, 1-4-chlorophenyl,1-p-chlorophenyl cyclopropanecarboxylic acid,acmc-209oos,1-4-chlorophenyl cyclopropanecarboxylicacid,1-4chloro-phenyl-cyclopropylcarboxylic acid,1-4-chloro-phenyl-cyclopropylcarboxylic acid
98606
1-(4-chlorophenyl)cyclopropane-1-carboxylic acid
C1CC1(C2=CC=C(C=C2)Cl)C(=O)O
Quantity 10g
Color White
Packaging Glass bottle
Melting Point 152.0°C to 155.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClC6H4C3H4CO2H
Formula Weight 196.63
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopropanecarboxylic acid, 99%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 277-105-5

TSCA : TSCA