Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol, 98%, Thermo Scientific™

Catalog No. AAH6473703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6473703 1 g
AAH6473706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6473703 Supplier Thermo Scientific Chemicals Supplier No. H6473703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133902-66-6
Molecular Formula C9H8ClN3O
Molecular Weight (g/mol) 209.633
MDL Number MFCD00140928
InChI Key CAHIFLPAMJOAGI-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl triazol-4-yl methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol, pubchem13003, chlorophenyltriazolylmethanol, 1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol
PubChem CID 2763878
IUPAC Name [1-(4-chlorophenyl)triazol-4-yl]methanol
SMILES C1=CC(=CC=C1N2C=C(N=N2)CO)Cl

Specifications

Melting Point 141°C to 145°C
Quantity 1 g
Beilstein 4250389
Formula Weight 209.63
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.