Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-3-buten-1-ol, 97%

Catalog No. AAH5338106
Click to view available options
Quantity:
1 g
5 g

CAS: 14506-33-3 | C10H11ClO | 182.647 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14506-33-3
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD03427246
InChI Key GJODRJDLQVBTMF-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-3-buten-1-ol, 1-4-chlorophenyl but-3-en-1-ol, benzenemethanol,4-chloro-a-2-propen-1-yl, 4-4-chlorophenyl-1-buten-4-ol, 1-4-chloro-phenyl-but-3-en-1-ol, acmc-20apb1, 4-chloro-alpha-allylbenzyl alcohol, 4-4-chlorophenyl but-1-en-4-ol, 1-4-chlorophenyl-but-3-en-1-ol
PubChem CID 2757771
IUPAC Name 1-(4-chlorophenyl)but-3-en-1-ol
SMILES C=CCC(C1=CC=C(C=C1)Cl)O

Specifications

Density 1.137
Boiling Point 140°C (2 mmHg)
Quantity 5 g
Refractive Index 1.544
Formula Weight 182.65
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-3-buten-1-ol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.