Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-chlorophenyl)ethanol, 98%, Thermo Scientific™

Catalog No. p-4258902
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 3391-10-4
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
InChI Key MVOSNPUNXINWAD-UHFFFAOYNA-N
Synonym boc-gly-osu, boc-glycine n-hydroxysuccinimide ester, boc-glycine-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2-tert-butoxycarbonyl amino acetate, tert-butyl 2-2,5-dioxo-1-pyrrolidinyl oxy-2-oxoethyl carbamate, bocglyosu, 2,5-dioxopyrrolidin-1-yl n-tert-butoxycarbonyl glycinate
PubChem CID 76926
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CC(O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 136°C (2.7 kPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 1.17
Solubility Information Solubility in water: in water:(20°C) insoluble
Formula Weight 156.61
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(4-chlorophenyl)ethanol
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-223-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.