Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanediol, 99%, mixture of cis and trans

Catalog No. p-2689466
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle

CAS: 556-48-9 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-48-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00001448,MFCD00063612,MFCD00075462
InChI Key VKONPUDBRVKQLM-UHFFFAOYSA-N
Synonym 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane
PubChem CID 11162
IUPAC Name cyclohexane-1,4-diol
SMILES OC1CCC(O)CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 98°C to 102°C
Boiling Point 150°C (20.0 mmHg)
Flash Point 65°C
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H10(OH)2
Quantity 25 g
Beilstein 06, IV, 5209
Solubility Information Solubility in water: soluble
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 1, 4-Cyclohexanediol, 99%
Show More Show Less
EINECSNumber 209-126-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.