Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanedione, 98%

Catalog No. p-4510281
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 637-88-7 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-88-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
InChI Key DCZFGQYXRKMVFG-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
PubChem CID 12511
ChEBI CHEBI:28286
IUPAC Name cyclohexane-1,4-dione
SMILES C1CC(=O)CCC1=O

Specifications

Melting Point 76.0°C to 80.0°C
Color Beige or Yellow
Boiling Point 112°C (26.0 hPa)
Flash Point 132°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H8(=O)2
Quantity 10 g
Beilstein 07,556
Fieser 01,178
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol
Formula Weight 112.13
Percent Purity 98%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 1, 4-Cyclohexanedione
Show More Show Less
EINECSNumber 211-306-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.