Learn More
1,4-Cyclohexanedione monoethylene acetal, 97%
Description
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Cyclohexanedione is used in the preparation of series of potent analgesic compounds. 1,4-Cyclohexanedione is also used as a building block in the synthesis of tritium labelled probes for the autoradiography study of the dopamine reuptake complex.
Solubility
Soluble in chloroform & methanol.
Notes
Moisture Sensitive. Store away from moisture and oxidizing agents. Incompatible with strong oxidizing agents and strong acids. Ensure adequate ventilation during use.

Specifications
Specifications
Melting Point | 72°C to 75°C |
Quantity | 25 g |
Beilstein | 117631 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform & methanol. |
Formula Weight | 156.18 |
Percent Purity | 97% |
Chemical Name or Material | 1,4-Cyclohexanedione monoethylene acetal |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 000-000-0 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.