Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibromobutane, 98+%

Catalog No. p-7081621
Click to view available options
Quantity:
10 g
100 g
500 g

CAS: 110-52-1 | C4H8Br2 | 215.916 g/mol

1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-52-1
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.916
MDL Number MFCD00000261
InChI Key ULTHEAFYOOPTTB-UHFFFAOYSA-N
Synonym tetramethylene dibromide, butane, 1,4-dibromo, 1,4-butylene bromide, tetramethylene bromide, 1,4-dibrombutan, tetramethylenebromide, alpha,omega-dibromobutane, unii-g49phr6jfu, 1,4-dibrombutan german, 1,4-dibromo-butane
PubChem CID 8056
IUPAC Name 1,4-dibromobutane
SMILES C(CCBr)CBr

Specifications

Melting Point -20°C
Density 1.82
Boiling Point 194°C to 196°C
Flash Point 131°C (267°F)
Refractive Index 1.5186
Linear Formula Br(CH2)4Br
Quantity 10 g
Beilstein 1071199
Solubility Information Immiscible with water.
Formula Weight 215.93
Percent Purity ≥98%
Chemical Name or Material 1,4-Dibromobutane
Show More Show Less
Hazard Category H301-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-775-5
RTECSNumber EJ7565000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.