Learn More
1,4-Dichloro-2-nitrobenzene, 98%
Description
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.
Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.
Notes
Incompatible with strong oxidizing agents, strong bases.

Specifications
Specifications
Melting Point | 53°C to 57°C |
Density | 1.6 |
Boiling Point | 266°C to 269°C |
Flash Point | 125°C (257°F) |
Quantity | 2500 g |
UN Number | UN3077 |
Beilstein | 778109 |
Solubility Information | Soluble in water,ethanol,ether,benzene,carbon disulfide. Slightly soluble in carbon tetrachloride. |
Formula Weight | 192 |
Percent Purity | 98% |
Show More |
Safety and Handling
Hazard Category | H302 |
Hazard Statement | GHS H Statement H341-H302-H319 Suspected of causing genetic defects. Harmful if swallowed. Causes serious eye irritation. |
Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 201-923-3 |
RTECSNumber | CZ5260000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.