Learn More
1,4-Dichlorobenzene, 99+%
Description
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Dichlorobenzene, is used as moth repellant, air fresheners and toilet blocks and general deodorant for garbage and restrom as well as to control fruit borers and ants, may be applied to tobacco seed beds for blue mold control. It is also used as a pharmaceutical intermediate. 1,4-dichlorobenzene is as a used as a minor processing aid in the production of grinding wheels.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, aluminum and its alloys, some plastics.
Specifications
Specifications
| Melting Point | 52°C to 56°C |
| Density | 1.248 |
| Boiling Point | 173°C to 174°C |
| Flash Point | 65°C (149°F) |
| Odor | Strong |
| Quantity | 2500 g |
| UN Number | UN3077 |
| Beilstein | 1680023 |
| Merck Index | 14,3057 |
| Solubility Information | It is insoluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H319-H350 |
| Hazard Statement | GHS H Statement H351-H302-H332-H319 Suspected of causing cancer. Harmful if swallowed. Harmful if inhaled. Causes serious eye irritation. |
| Precautionary Statement | P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 203-400-5 |
| RTECSNumber | CZ4550000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.