Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dihydro-4-oxoquinoline-3-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. D48811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48811G 1 g
D48815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48811G Supplier TCI America Supplier No. D48811G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13721-01-2
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00498984
InChI Key ILNJBIQQAIIMEY-UHFFFAOYSA-N
PubChem CID 220875
IUPAC Name 4-oxo-1H-quinoline-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=O)C(=CN2)C(=O)O

Specifications

Melting Point 265°C
Color White-Yellow
Quantity 1 g
Formula Weight 189.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Dihydro-4-oxoquinoline-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.