Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diiodo-2,3,5,6-tetramethylbenzene 95.0+%, TCI America™
SDP

Catalog No. D202825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D202825G 25 g
D20285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D202825G Supplier TCI America Supplier No. D202825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3268-21-1
Molecular Formula C10H12I2
Molecular Weight (g/mol) 386.02
MDL Number MFCD00142554
InChI Key YSKCEXICRGEWDI-UHFFFAOYSA-N
Synonym 3,6-Diiododurene
PubChem CID 5182526
IUPAC Name 1,4-diiodo-2,3,5,6-tetramethylbenzene
SMILES CC1=C(C)C(I)=C(C)C(C)=C1I

Specifications

Melting Point 140°C
Color White
Quantity 25 g
Formula Weight 386.01
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diiodo-2,3,5,6-tetramethylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.