Learn More
1,4-Diisopropylbenzene, 98%, Thermo Scientific Chemicals
Description
1,4-Diisopropylbenzene is used as an intermediate for the synthesis of stabilizers, polymers, synthetic lubricants and hydroperoxides. It is also used in the trivalent actinide-lanthanide separation by phosphorus reagent extraction from aqueous komplexes (TALSPEAK) process for separating trivalent lanthanides from trivalent actinide cations. It finds application in nuclear fuel reprocessing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Diisopropylbenzene is used as an intermediate for the synthesis of stabilizers, polymers, synthetic lubricants and hydroperoxides. It is also used in the trivalent actinide-lanthanide separation by phosphorus reagent extraction from aqueous komplexes (TALSPEAK) process for separating trivalent lanthanides from trivalent actinide cations. It finds application in nuclear fuel reprocessing.
Solubility
Soluble in alcohol, ether, acetone and benzene. Insoluble in water.
Notes
Incompatible with oxidizing agents and strong oxidizing agents.
Specifications
Melting Point | -17°C |
Density | 0.857 |
Boiling Point | 210°C |
Flash Point | 76°C (170°F) |
Odor | Aromatic |
Refractive Index | 1.489 |
Quantity | 25 g |
Beilstein | 1854739 |
Solubility Information | Soluble in alcohol,ether,acetone and benzene. Insoluble in water. |
Formula Weight | 162.27 |
Show More |
Safety and Handling
Hazard Category | H227-H315-H319-H335 |
Hazard Statement | GHS H Statement H227 Combustible liquid. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 202-826-9 |
RTECSNumber | CZ6360000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.