Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diisopropylbenzene, 99%

Catalog No. p-113216
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 100-18-5 | C12H18 | 162.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-18-5
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00008892
InChI Key SPPWGCYEYAMHDT-UHFFFAOYSA-N
Synonym 1,4-diisopropylbenzene, p-diisopropylbenzene, benzene, p-diisopropyl, benzene, 1,4-bis 1-methylethyl, para-diisopropylbenzene, p-diisopropylbenzol, 1,4-bis 1-methylethyl benzene, unii-5m62031nzp, 1,4-di propan-2-yl benzene, dsstox_cid_6652
PubChem CID 7486
SMILES CC(C)C1=CC=C(C=C1)C(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -17.0°C
Density 0.8600g/mL
Boiling Point 203.0°C
Flash Point 76°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4890 to 1.4910
Linear Formula C6H4[CH(CH3)2]2
Quantity 5 g
Beilstein 05,II,339
Specific Gravity 0.86
Solubility Information Solubility in water: insoluble. Other solubilities: %iscible with alcohol, ether, benzene, acetone and, ccl4
IUPAC Name 1,4-bis(propan-2-yl)benzene
Formula Weight 162.27
Percent Purity 99%
Chemical Name or Material 1,4-Diisopropylbenzene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 202-826-9
RTECSNumber CZ6360000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.